COM448687088 Combi-Blocks, Inc.

methyl 2-chloro-4-(4-chlorophenyl)benzoate

  • MDL number MFCD27578682
  • Cas Number : 1820716-21-9
  • Molecular Weight : 281.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2187

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2187
Availability details for
Quantity
  • Availability
  • Add to Cart