COM448687089 Combi-Blocks, Inc.

methyl 3-chloro-5-(thiophen-3-yl)benzoate

  • MDL number MFCD27578683
  • Cas Number : 177735-07-8
  • Molecular Weight : 252.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2189

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2189
Availability details for
Quantity
  • Availability
  • Add to Cart