COM448687090 Combi-Blocks, Inc.

5-Fluoro-3'-(trifluoromethyl)biphenyl-3-carboxylic acid methyl ester

  • MDL number MFCD18425056
  • Cas Number : 1261633-95-7
  • Molecular Weight : 298.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2190

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2190
Availability details for
Quantity
  • Availability
  • Add to Cart