COM448687091 Combi-Blocks, Inc.

methyl 2-{4-[3-(trifluoromethyl)phenyl]phenyl}acetate

  • MDL number MFCD27578684
  • Cas Number : 877607-36-8
  • Molecular Weight : 294.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2193

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2193
Availability details for
Quantity
  • Availability
  • Add to Cart