COM448687096 Combi-Blocks, Inc.

methyl 4-(3-fluorophenyl)-3-methylbenzoate

  • MDL number MFCD27578687
  • Cas Number : 1820706-80-6
  • Molecular Weight : 244.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2200

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2200
Availability details for
Quantity
  • Availability
  • Add to Cart