COM448687101 Combi-Blocks, Inc.

methyl 4-[3-(benzyloxy)phenyl]-2-chlorobenzoate

  • MDL number MFCD27578691
  • Cas Number : 1820712-00-2
  • Molecular Weight : 352.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2209

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2209
Availability details for
Quantity
  • Availability
  • Add to Cart