COM448687107 Combi-Blocks, Inc.

2-Fluoro-4'-(trifluoromethyl)biphenyl-4-carboxylic acid methyl ester

  • MDL number MFCD18425040
  • Cas Number : 1261480-17-4
  • Molecular Weight : 298.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2216

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2216
Availability details for
Quantity
  • Availability
  • Add to Cart