COM448687113 Combi-Blocks, Inc.

methyl 2-(3-fluoro-4-methylphenyl)benzoate

  • MDL number MFCD27578697
  • Cas Number : 138459-23-1
  • Molecular Weight : 244.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2225

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2225
Availability details for
Quantity
  • Availability
  • Add to Cart