COM448687123 Combi-Blocks, Inc.

methyl 3-chloro-5-(4-fluorophenyl)benzoate

  • MDL number MFCD27578706
  • Cas Number : 1820620-19-6
  • Molecular Weight : 264.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2237

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2237
Availability details for
Quantity
  • Availability
  • Add to Cart