COM448687126 Combi-Blocks, Inc.

Methyl 2-methyl-3-phenylbenzoate

  • MDL number MFCD18379547
  • Cas Number : 1329496-87-8
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2240

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2240
Availability details for
Quantity
  • Availability
  • Add to Cart