COM448687131 Combi-Blocks, Inc.

Methyl 3-(tert-butylsulfamoyl)benzoate

  • MDL number MFCD20044428
  • Cas Number : 1457311-34-0
  • Molecular Weight : 271.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2248

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2248
Availability details for
Quantity
  • Availability
  • Add to Cart