COM448687134 Combi-Blocks, Inc.

methyl 3-(3,5-dimethylphenyl)-2-methylbenzoate

  • MDL number MFCD27578713
  • Cas Number : 1820684-22-7
  • Molecular Weight : 254.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2252

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2252
Availability details for
Quantity
  • Availability
  • Add to Cart