COM448687139 Combi-Blocks, Inc.

Dimethyl 5-(trifluoromethyl)benzene-1,3-dicarboxylate

  • MDL number MFCD27578718
  • Cas Number : 868286-79-7
  • Molecular Weight : 262.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2257

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2257
Availability details for
Quantity
  • Availability
  • Add to Cart