COM448687146 Combi-Blocks, Inc.

methyl 3-(3-chlorophenyl)-4-fluorobenzoate

  • MDL number MFCD27578724
  • Cas Number : 1820740-85-9
  • Molecular Weight : 264.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2265

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2265
Availability details for
Quantity
  • Availability
  • Add to Cart