COM448687149 Combi-Blocks, Inc.

Methyl 2-hydroxy-3-methyl-2-phenylbutanoate

  • MDL number MFCD00048318
  • Cas Number : 72433-75-1
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2272

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2272
Availability details for
Quantity
  • Availability
  • Add to Cart