COM448687152 Combi-Blocks, Inc.

Methyl 3-(4-sulfamoylphenyl)propanoate

  • MDL number MFCD18839950
  • Cas Number : 928139-28-0
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2276

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2276
Availability details for
Quantity
  • Availability
  • Add to Cart