COM448687160 Combi-Blocks, Inc.

methyl 2-chloro-4-[3-fluoro-4-(methoxycarbonyl)phenyl]benzoate

  • MDL number MFCD27578734
  • Cas Number : 1820648-58-5
  • Molecular Weight : 322.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2287

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2287
Availability details for
Quantity
  • Availability
  • Add to Cart