COM448687162 Combi-Blocks, Inc.

Methyl 2-(3-fluorophenyl)benzoate

  • MDL number MFCD05979194
  • Cas Number : 80254-81-5
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2289

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2289
Availability details for
Quantity
  • Availability
  • Add to Cart