COM448687163 Combi-Blocks, Inc.

methyl 2-chloro-4-(2,4-difluorophenyl)benzoate

  • MDL number MFCD27578736
  • Cas Number : 1820705-05-2
  • Molecular Weight : 282.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2290

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2290
Availability details for
Quantity
  • Availability
  • Add to Cart