COM448687167 Combi-Blocks, Inc.

methyl 3-(2-fluoro-5-methylphenyl)-5-methoxybenzoate

  • MDL number MFCD27578739
  • Cas Number : 1820649-40-8
  • Molecular Weight : 274.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2297

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2297
Availability details for
Quantity
  • Availability
  • Add to Cart