COM448687168 Combi-Blocks, Inc.

methyl 2-[3-(2-methylphenyl)phenyl]acetate

  • MDL number MFCD27578740
  • Cas Number : 1820706-84-0
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2298

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2298
Availability details for
Quantity
  • Availability
  • Add to Cart