COM448687172 Combi-Blocks, Inc.

methyl 2-[3-(2-chloro-4-methoxyphenyl)phenyl]acetate

  • MDL number MFCD27578743
  • Cas Number : 1820717-68-7
  • Molecular Weight : 290.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2302

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2302
Availability details for
Quantity
  • Availability
  • Add to Cart