COM448687173 Combi-Blocks, Inc.

methyl 4-(2,4-dimethylphenyl)pyridine-2-carboxylate

  • MDL number MFCD27578744
  • Cas Number : 1820649-68-0
  • Molecular Weight : 241.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2303

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2303
Availability details for
Quantity
  • Availability
  • Add to Cart