COM448687179 Combi-Blocks, Inc.

methyl 3-[2-(dimethylsulfamoyl)phenyl]-4-fluorobenzoate

  • MDL number MFCD27578750
  • Cas Number : 1820717-72-3
  • Molecular Weight : 337.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2310

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2310
Availability details for
Quantity
  • Availability
  • Add to Cart