COM448687180 Combi-Blocks, Inc.

Fmoc-Arg(pbf)-OMe

  • MDL number MFCD27578751
  • Cas Number : 813452-47-0
  • Molecular Weight : 662.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2311

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2311
Availability details for
Quantity
  • Availability
  • Add to Cart