COM448687184 Combi-Blocks, Inc.

methyl 3-chloro-5-(2,3-dichlorophenyl)benzoate

  • MDL number MFCD27578753
  • Cas Number : 1820703-24-9
  • Molecular Weight : 315.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2316

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2316
Availability details for
Quantity
  • Availability
  • Add to Cart