COM448687195 Combi-Blocks, Inc.

1,4-dimethyl 2,5-diphenylbenzene-1,4-dicarboxylate

  • MDL number MFCD27578763
  • Cas Number : 16308-63-7
  • Molecular Weight : 346.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2332

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2332
Availability details for
Quantity
  • Availability
  • Add to Cart