COM448687205 Combi-Blocks, Inc.

methyl 4-acetamidopyridine-2-carboxylate

  • MDL number MFCD27578769
  • Cas Number : 1820716-80-0
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2346

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2346
Availability details for
Quantity
  • Availability
  • Add to Cart