COM448687206 Combi-Blocks, Inc.

Methyl (2-methyl-1h-benzimidazol-1-yl)acetate

  • MDL number MFCD05997543
  • Cas Number : 2033-54-7
  • Molecular Weight : 204.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2347

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2347
Availability details for
Quantity
  • Availability
  • Add to Cart