COM448687207 Combi-Blocks, Inc.

methyl 2-[4-(4-methylphenyl)phenyl]acetate

  • MDL number MFCD27578770
  • Cas Number : 1437789-26-8
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2348

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2348
Availability details for
Quantity
  • Availability
  • Add to Cart