COM448687209 Combi-Blocks, Inc.

Methyl 4-(6-methoxy-3-pyridinyl)benzoate

  • MDL number MFCD06801888
  • Cas Number : 406234-25-1
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2351

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2351
Availability details for
Quantity
  • Availability
  • Add to Cart