COM448687211 Combi-Blocks, Inc.

methyl 2-(3,5-dimethylphenyl)benzoate

  • MDL number MFCD27578772
  • Cas Number : 1528793-40-9
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2354

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2354
Availability details for
Quantity
  • Availability
  • Add to Cart