COM448687212 Combi-Blocks, Inc.

methyl 2-{4-[(methoxycarbonyl)amino]phenyl}benzoate

  • MDL number MFCD27578773
  • Cas Number : 1820716-94-6
  • Molecular Weight : 285.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2356

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2356
Availability details for
Quantity
  • Availability
  • Add to Cart