COM448687219 Combi-Blocks, Inc.

methyl 2-fluoro-5-[3-(methoxycarbonyl)-5-nitrophenyl]benzoate

  • MDL number MFCD27578778
  • Cas Number : 1820620-22-1
  • Molecular Weight : 333.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2367

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2367
Availability details for
Quantity
  • Availability
  • Add to Cart