COM448687228 Combi-Blocks, Inc.

Methyl 3-(4-fluorobenzenesulfonamido)propanoate

  • MDL number MFCD13223606
  • Cas Number : 208121-68-0
  • Molecular Weight : 261.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2377

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2377
Availability details for
Quantity
  • Availability
  • Add to Cart