COM448687235 Combi-Blocks, Inc.

methyl 2-{4-[4-(ethylcarbamoyl)-3-fluorophenyl]phenyl}acetate

  • MDL number MFCD27578789
  • Cas Number : 1820640-21-8
  • Molecular Weight : 315.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2384

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2384
Availability details for
Quantity
  • Availability
  • Add to Cart