COM448687238 Combi-Blocks, Inc.

methyl 4-(3,4-difluorophenyl)-2-fluorobenzoate

  • MDL number MFCD27578792
  • Cas Number : 1820640-08-1
  • Molecular Weight : 266.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2387

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2387
Availability details for
Quantity
  • Availability
  • Add to Cart