COM448687245 Combi-Blocks, Inc.

methyl 4-(2-acetylphenyl)-2-fluorobenzoate

  • MDL number MFCD27578799
  • Cas Number : 1820648-54-1
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2398

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2398
Availability details for
Quantity
  • Availability
  • Add to Cart