COM448687250 Combi-Blocks, Inc.

methyl 3-[3-(benzyloxy)phenyl]-5-nitrobenzoate

  • MDL number MFCD27578803
  • Cas Number : 1820650-14-3
  • Molecular Weight : 363.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2406

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2406
Availability details for
Quantity
  • Availability
  • Add to Cart