COM448687252 Combi-Blocks, Inc.

Methyl 2'-chloro-1,1'-biphenyl-4-carboxylate

  • MDL number MFCD05981681
  • Cas Number : 89900-96-9
  • Molecular Weight : 246.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2408

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2408
Availability details for
Quantity
  • Availability
  • Add to Cart