COM448687258 Combi-Blocks, Inc.

Methyl 4-(acetamidomethyl)benzoate

  • MDL number MFCD10653289
  • Cas Number : 20686-40-2
  • Molecular Weight : 207.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2418

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2418
Availability details for
Quantity
  • Availability
  • Add to Cart