COM448687259 Combi-Blocks, Inc.

methyl 3-fluoro-4-(4-methylphenyl)benzoate

  • MDL number MFCD27578809
  • Cas Number : 1820674-27-8
  • Molecular Weight : 244.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2419

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2419
Availability details for
Quantity
  • Availability
  • Add to Cart