COM448687263 Combi-Blocks, Inc.

methyl 2-chloro-4-[4-(methylsulfanyl)phenyl]benzoate

  • MDL number MFCD27578813
  • Cas Number : 1820739-54-5
  • Molecular Weight : 292.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2423

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2423
Availability details for
Quantity
  • Availability
  • Add to Cart