COM448687264 Combi-Blocks, Inc.

methyl 3-fluoro-4-(4-fluoro-3-methylphenyl)benzoate

  • MDL number MFCD27578814
  • Cas Number : 1820717-95-0
  • Molecular Weight : 262.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2425

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2425
Availability details for
Quantity
  • Availability
  • Add to Cart