COM448687268 Combi-Blocks, Inc.

methyl 3-(2-chloro-4-methylphenyl)-5-nitrobenzoate

  • MDL number MFCD27578818
  • Cas Number : 1820614-02-5
  • Molecular Weight : 305.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2429

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2429
Availability details for
Quantity
  • Availability
  • Add to Cart