COM448687269 Combi-Blocks, Inc.

methyl 2-(2,6-dichloro-3-fluorophenyl)-2-oxoacetate

  • MDL number MFCD27578819
  • Cas Number : 1820739-90-9
  • Molecular Weight : 251.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2430

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2430
Availability details for
Quantity
  • Availability
  • Add to Cart