COM448687275 Combi-Blocks, Inc.

methyl 6-(4-cyanophenyl)pyridine-2-carboxylate

  • MDL number MFCD27578825
  • Cas Number : 1820607-47-3
  • Molecular Weight : 238.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2437

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2437
Availability details for
Quantity
  • Availability
  • Add to Cart