COM448687276 Combi-Blocks, Inc.

Methyl 6-thien-2-ylnicotinate

  • MDL number MFCD08690297
  • Cas Number : 179408-53-8
  • Molecular Weight : 219.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2438

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2438
Availability details for
Quantity
  • Availability
  • Add to Cart