COM448687277 Combi-Blocks, Inc.

methyl 3-methoxy-5-(2-methylphenyl)benzoate

  • MDL number MFCD27578826
  • Cas Number : 1820675-15-7
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2439

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2439
Availability details for
Quantity
  • Availability
  • Add to Cart