COM448687280 Combi-Blocks, Inc.

methyl 3-(3-bromo-4-methoxyphenyl)benzoate

  • MDL number MFCD27578829
  • Cas Number : 1820608-00-1
  • Molecular Weight : 321.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2442

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2442
Availability details for
Quantity
  • Availability
  • Add to Cart